Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000298596.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399162.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399162.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399162.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399162.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399162.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399162.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
ENST00000399169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_001130161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007871.hg.1 |
NM_152709.4 |
Refseq |