Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000406.hg.1 |
ENST00000319006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000406.hg.1 |
ENST00000528097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000406.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000406.hg.1 |
TC11000027.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000407.hg.1 |
ENST00000319006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000407.hg.1 |
ENST00000528097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000407.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000407.hg.1 |
TC11000027.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144821.hg.1 |
TC1100006487.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144824.hg.1 |
TC1100006487.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000319006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000528097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000530666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000532202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000532685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
ENST00000533796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006487.hg.1 |
NM_006755.1 |
Refseq |