Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000305762.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000322635.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000322680.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000351293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000372765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000394762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000423381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000433289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000441192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000472912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000474131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_001204492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_001222.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172170.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000305762.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000322635.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000322680.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000351293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000372765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000394762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000423381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000433289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000441192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000472912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
ENST00000474131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_001204492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_001222.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172170.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
NM_172173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175353.hg.1 |
TC1000011040.hg.1 |
Clariom D TC |