Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009867.hg.1 |
TC02000621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009867.hg.1 |
ENST00000264257.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009867.hg.1 |
ENST00000441515.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009867.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009867.hg.1 |
ENST00000539491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009867.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160550.hg.1 |
TC0200008669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000264257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000421464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000441515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
ENST00000481806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008669.hg.1 |
NM_003854.3 |
Refseq |