Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
TC02000714.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
ENST00000263326.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
ENST00000349806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
ENST00000352179.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
ENST00000353225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011085.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
TC02000714.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
ENST00000263326.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
ENST00000311328.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
ENST00000349806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
ENST00000352179.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
ENST00000353225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011089.hg.1 |
NM_173205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
TC02000714.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
ENST00000263326.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
ENST00000349806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
ENST00000352179.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
ENST00000353225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011084.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000263326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000311328.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000349806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000352179.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000353225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000263326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000311328.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000349806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000352179.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
ENST00000353225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
NM_173205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162448.hg.1 |
TC0200008942.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000263326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000349806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000352179.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000353225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000263326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000349806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000352179.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
ENST00000353225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162442.hg.1 |
TC0200008942.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000263326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000349806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000352179.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000353225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000263326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000349806.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000352179.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
ENST00000353225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_014439.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_173202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_173203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
NM_173204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162444.hg.1 |
TC0200008942.hg.1 |
Clariom D TC |