Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007658.hg.1 |
TC05000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007658.hg.1 |
ENST00000357872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007658.hg.1 |
ENST00000379578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007658.hg.1 |
ENST00000512413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007658.hg.1 |
NM_173800.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007665.hg.1 |
TC05000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007665.hg.1 |
ENST00000357872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007665.hg.1 |
ENST00000379578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007665.hg.1 |
ENST00000512413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007665.hg.1 |
NM_173800.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007657.hg.1 |
TC05000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007657.hg.1 |
ENST00000357872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007657.hg.1 |
ENST00000379578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007657.hg.1 |
ENST00000512413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007657.hg.1 |
NM_173800.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007663.hg.1 |
TC05000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007663.hg.1 |
ENST00000357872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007663.hg.1 |
ENST00000379578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007663.hg.1 |
ENST00000512413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007663.hg.1 |
NM_173800.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007659.hg.1 |
TC05000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007659.hg.1 |
ENST00000357872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007659.hg.1 |
ENST00000379578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007659.hg.1 |
ENST00000512413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007659.hg.1 |
NM_173800.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000357872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000395528.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000503329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000512413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
NM_173800.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000357872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000395528.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000503329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
ENST00000512413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
NM_173800.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193852.hg.1 |
TC0500013215.hg.1 |
Clariom D TC |