Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027119.hg.1 |
ENST00000355329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027119.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027119.hg.1 |
TC12002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027120.hg.1 |
ENST00000355329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027120.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027120.hg.1 |
TC12002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027121.hg.1 |
ENST00000355329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027121.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027121.hg.1 |
TC12002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027123.hg.1 |
ENST00000355329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027123.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027123.hg.1 |
TC12002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027122.hg.1 |
ENST00000355329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027122.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027122.hg.1 |
TC12002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195343.hg.1 |
TC1200012210.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012210.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195345.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195345.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195345.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195345.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195345.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195345.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195345.hg.1 |
TC1200012210.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195341.hg.1 |
TC1200012210.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195344.hg.1 |
TC1200012210.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200195340.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195340.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195340.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195340.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195340.hg.1 |
TC1200012210.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
ENST00000355329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
ENST00000392462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
ENST00000542364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
NM_173855.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195342.hg.1 |
TC1200012210.hg.1 |
Clariom D TC |