Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004244.hg.1 |
TC07000274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004244.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004244.hg.1 |
ENST00000447123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004244.hg.1 |
ENST00000496112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004244.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004243.hg.1 |
TC07000274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004243.hg.1 |
ENST00000447123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004243.hg.1 |
ENST00000496112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004243.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004242.hg.1 |
TC07000274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004242.hg.1 |
ENST00000496112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004242.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000478411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000478411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000478411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013735.hg.1 |
ENST00000434104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013735.hg.1 |
ENST00000434104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004245.hg.1 |
TC07000274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004245.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004245.hg.1 |
ENST00000447123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004245.hg.1 |
ENST00000496112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004245.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151197.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151197.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151197.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151197.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151197.hg.1 |
TC0700007374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151201.hg.1 |
TC0700007374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013735.hg.1 |
ENST00000434104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013735.hg.1 |
ENST00000434104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151198.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151198.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151198.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151198.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151198.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151198.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151198.hg.1 |
TC0700007374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151200.hg.1 |
TC0700007374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000421621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000424864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000463014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000478411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000478411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000478411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007374.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
ENST00000223369.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
ENST00000447123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
ENST00000496112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
NM_006555.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151199.hg.1 |
TC0700007374.hg.1 |
Clariom D TC |