Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188361.hg.1 |
TC0500012662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012662.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188362.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188362.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188362.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188362.hg.1 |
ENST00000408953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188362.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188362.hg.1 |
NM_022090.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188362.hg.1 |
TC0500012662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500188360.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188360.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188360.hg.1 |
ENST00000523213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188360.hg.1 |
NM_001303251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188360.hg.1 |
TC0500012662.hg.1 |
Clariom D TC |