Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054704.hg.1 |
TC01003528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054704.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054704.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054704.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000476568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000476568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000476568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224878.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224878.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224878.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224878.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224878.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224878.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224878.hg.1 |
TC0100016437.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000367714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000466087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000476568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000476568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000476568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
ENST00000479367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016437.hg.1 |
NM_178527.3 |
Refseq |