Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000494418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000494418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
ENST00000494418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175820.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175820.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175820.hg.1 |
TC0100010331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175826.hg.1 |
TC0100010331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000368011.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000392192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000469647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000475934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000490724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000494418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000494418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
ENST00000494418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_001007255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NM_152366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010331.hg.1 |
NR_033386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175816.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175816.hg.1 |
ENST00000471613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175816.hg.1 |
TC0100010331.hg.1 |
Clariom D TC |