Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
TC2000007197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149389.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374809.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
ENST00000451665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007197.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149391.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149391.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149391.hg.1 |
NM_021202.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149391.hg.1 |
TC2000007197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149391.hg.1 |
ENST00000374810.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149391.hg.1 |
ENST00000414082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149391.hg.1 |
NM_021202.2 |
Refseq |