Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019831.hg.1 |
TC02001299.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019831.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019831.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019831.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019827.hg.1 |
TC02001299.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019827.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019827.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019827.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000494447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010794.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176669.hg.1 |
TC0200010794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000289388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000480532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000481940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
ENST00000495773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
NM_198559.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176665.hg.1 |
TC0200010794.hg.1 |
Clariom D TC |