Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_175900_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8000704 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8000705 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8003604 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8003603 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8003602 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8000703 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8003601 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8000702 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1559584_PM_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735767_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735768_PM_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735766_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735766_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735767_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735768_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014446.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014446.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014446.hg.1 |
TC16001008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009937.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176422.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176422.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176422.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176422.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176422.hg.1 |
TC1600009937.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600176423.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176423.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176423.hg.1 |
ENST00000329410.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176423.hg.1 |
NM_175900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176423.hg.1 |
TC1600009937.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
1559584_a_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1559584_PM_a_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1559584_a_at3 |
- |
- |