Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008684.hg.1 |
ENST00000579042.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000384602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000496648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000496648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
ENST00000496648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008684.hg.1 |
ENST00000579042.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000262848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000384602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000425240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000462736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000496648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000496648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
ENST00000496648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008945.hg.1 |
NM_005044.4 |
Refseq |