Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_012404_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955118 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7982532 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7955117 |
- |
- |
Affymetrix Human Genome U219 Array |
11738964_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11738964_at |
- |
- |
Affymetrix Human Genome U219 Array |
11738965_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11738965_PM_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007521.hg.1 |
ENST00000266594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007521.hg.1 |
ENST00000266594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007521.hg.1 |
NM_012404.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004387.hg.1 |
ENST00000266594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004387.hg.1 |
NM_012404.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004387.hg.1 |
TC12000350.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200007521.hg.1 |
ENST00000266594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007521.hg.1 |
ENST00000266594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007521.hg.1 |
NM_012404.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200152856.hg.1 |
ENST00000266594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200152856.hg.1 |
NM_012404.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200152856.hg.1 |
ENST00000266594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200152856.hg.1 |
NM_012404.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200152856.hg.1 |
TC1200007521.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
208571_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
208571_PM_at |
- |
- |
Human Genome U133A 2.0 Array |
208571_PM_at |
- |
- |
Human Genome U133A 2.0 Array |
208571_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_012404_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw585199 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208571_at7 |
- |
- |
Human Genome U133 Set |
208571_PM_at |
- |
- |
Human Genome U133 Set |
208571_at |
- |
- |