Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019117.hg.1 |
TC01001248.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019117.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019117.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010020.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010020.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010020.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010020.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010020.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010020.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010020.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010020.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010020.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010020.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010020.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010020.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172624.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172624.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172624.hg.1 |
ENST00000307098.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172624.hg.1 |
NM_003125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172624.hg.1 |
TC0100010020.hg.1 |
Clariom D TC |