Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007664.hg.1 |
ENST00000374940.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000343139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000374949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000395363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000422863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000460633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000482745.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
ENST00000496318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007657.hg.1 |
NM_002122.3 |
Refseq |