Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015890.hg.1 |
TC09001276.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015890.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015890.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000340717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010565.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900172030.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900172030.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900172030.hg.1 |
ENST00000376447.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900172030.hg.1 |
NM_152573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900172030.hg.1 |
TC0900010565.hg.1 |
Clariom D TC |