Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008659.hg.1 |
ENST00000412537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007211.hg.1 |
ENST00000420036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013903.hg.1 |
TC05000956.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013903.hg.1 |
ENST00000519374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013903.hg.1 |
ENST00000519911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013903.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166368.hg.1 |
TC0500009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166367.hg.1 |
TC0500009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166365.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166365.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166365.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166365.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166365.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166365.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166365.hg.1 |
TC0500009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007211.hg.1 |
ENST00000420036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166369.hg.1 |
TC0500009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008659.hg.1 |
ENST00000412537.1 |
Ensembl |