Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193999.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193999.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193999.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193999.hg.1 |
TC1900011195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193999.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193999.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193999.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |