Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2BFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164416.1 | NP_001157888.1 | 3 | 712 | 85 | |
GenBank mRNA | AK093522.1 | - | 3 | 712 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589263 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589301 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589306 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589262 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859277 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859280 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859273 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383630 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859274 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859269 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383635 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589330 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383607 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589338 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589282 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589322 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859295 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589325 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859300 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859275 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589331 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859263 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589280 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859264 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589342 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383626 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589279 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859296 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383597 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11859287 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589268 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32589339 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383585 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3222689 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6935 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3222689 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3222689 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859275 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859269 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589285 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859269 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589285 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859275 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589268 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 4147 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P17247099 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P763762 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8169082 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8169081 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8169073 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8169080 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 234899_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006783.hg.1 | TC0X000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006783.hg.1 | ENST00000243297.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006783.hg.1 | ENST00000355016.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006783.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006784.hg.1 | TC0X000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006784.hg.1 | ENST00000243297.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006784.hg.1 | ENST00000355016.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006784.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006782.hg.1 | TC0X000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006782.hg.1 | ENST00000243297.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006782.hg.1 | ENST00000355016.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006782.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006781.hg.1 | TC0X000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006781.hg.1 | ENST00000243297.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006781.hg.1 | ENST00000355016.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006781.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006785.hg.1 | TC0X000507.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006785.hg.1 | ENST00000243297.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006785.hg.1 | ENST00000355016.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006785.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | ENST00000417637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | ENST00000598335.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | ENST00000417637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | ENST00000598335.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154827.hg.1 | TC0X00008026.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00154829.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154829.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154829.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154829.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154829.hg.1 | TC0X00008026.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00154830.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154830.hg.1 | ENST00000417637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154830.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154830.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154830.hg.1 | ENST00000417637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154830.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154830.hg.1 | TC0X00008026.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00154825.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154825.hg.1 | ENST00000598335.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154825.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154825.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154825.hg.1 | ENST00000598335.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154825.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154825.hg.1 | TC0X00008026.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00154824.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154824.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154824.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154824.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154824.hg.1 | TC0X00008026.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | ENST00000417637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | ENST00000598335.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | ENST00000355016.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | ENST00000417637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | ENST00000598335.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | NM_001164416.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00154826.hg.1 | TC0X00008026.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 234899_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 234899_at | - | - |
Human Genome U133 Plus 2.0 Array | 230664_at | - | - |
Human Genome U133 Plus 2.0 Array | 230664_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234899_at7 | - | - |
Human Genome U95 Set | 64216_at | - | - |
Human Genome U95 Set | 64216_PM_at | - | - |
Human Genome U133 Set | 234899_at | - | - |
Human Genome U133 Set | 234899_PM_at | - | - |
Human Genome U133 Set | 230664_at | - | - |
Human Genome U133 Set | 230664_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041352 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041352 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 211676 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1675025 | AK093522.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1675025 | NM_001164416.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1675025 | AK093522.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1675025 | NM_001164416.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1675025 | AK093522.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1675025 | NM_001164416.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1675025 | AK093522.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1675025 | NM_001164416.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1675025 | AK093522.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1675025 | NM_001164416.1 | human_transcriptome |