Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000467495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000467495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
ENST00000467495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187014.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187014.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187014.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187014.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187014.hg.1 |
TC0100011797.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000366789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000467495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000467495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
ENST00000467495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011797.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187015.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187015.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187015.hg.1 |
ENST00000366788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187015.hg.1 |
NM_001003665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187015.hg.1 |
TC0100011797.hg.1 |
Clariom D TC |