Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018734.hg.1 |
TC06004125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018734.hg.1 |
ENST00000438763.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018734.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003519.hg.1 |
TC6_mcf_hap5000224.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003519.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003454.hg.1 |
TC6_ssto_hap7000212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003454.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003629.hg.1 |
TC6_dbb_hap3000241.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003629.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003522.hg.1 |
TC6_mcf_hap5000224.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003522.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003879.hg.1 |
TC6_cox_hap2000256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003879.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018735.hg.1 |
TC06004125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018735.hg.1 |
ENST00000438763.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018735.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003882.hg.1 |
TC6_cox_hap2000256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003882.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018738.hg.1 |
TC06004125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018738.hg.1 |
ENST00000438763.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018738.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001685.hg.1 |
TC6_apd_hap1000129.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001685.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003878.hg.1 |
TC6_cox_hap2000256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003878.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003451.hg.1 |
TC6_ssto_hap7000212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003451.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003630.hg.1 |
TC6_dbb_hap3000241.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003630.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003633.hg.1 |
TC6_dbb_hap3000241.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003633.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003518.hg.1 |
TC6_mcf_hap5000224.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003518.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001684.hg.1 |
TC6_apd_hap1000129.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001684.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001688.hg.1 |
TC6_apd_hap1000129.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001688.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003450.hg.1 |
TC6_ssto_hap7000212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003450.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018733.hg.1 |
TC06004125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018733.hg.1 |
ENST00000438763.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018733.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201491.hg.1 |
TC0600014274.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000447394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014274.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201488.hg.1 |
TC0600014274.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
ENST00000438763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
ENST00000475235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
ENST00000488325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
NM_002120.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201489.hg.1 |
TC0600014274.hg.1 |
Clariom D TC |