Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005718.hg.1 |
ENST00000216044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005718.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005718.hg.1 |
TC22000304.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005721.hg.1 |
ENST00000216044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005721.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005721.hg.1 |
TC22000304.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005722.hg.1 |
ENST00000216044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005722.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005722.hg.1 |
TC22000304.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009267.hg.1 |
ENST00000487538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009267.hg.1 |
ENST00000487538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009267.hg.1 |
ENST00000487538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009267.hg.1 |
ENST00000489527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009267.hg.1 |
ENST00000489527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005720.hg.1 |
ENST00000216044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005720.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005720.hg.1 |
TC22000304.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200168833.hg.1 |
TC2200009266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168833.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168833.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168833.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168833.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168833.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168833.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009267.hg.1 |
ENST00000487538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009267.hg.1 |
ENST00000487538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009267.hg.1 |
ENST00000487538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009267.hg.1 |
ENST00000489527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009267.hg.1 |
ENST00000489527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168830.hg.1 |
TC2200009266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168830.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168830.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168830.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168830.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168830.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168830.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168831.hg.1 |
TC2200009266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168831.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168831.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168831.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168831.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168831.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168831.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168832.hg.1 |
TC2200009266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168832.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168832.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168832.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168832.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168832.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168832.hg.1 |
NM_004286.4 |
Refseq |