Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000501338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000502262.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013341.hg.1 |
ENST00000504967.2 |
Ensembl |