Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004991.hg.1 |
TC01006315.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004991.hg.1 |
ENST00000354177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004991.hg.1 |
ENST00000361547.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004991.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004991.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004991.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237494.hg.1 |
TC0100018200.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000354177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000361547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000374315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000494537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
ENST00000630065.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_020451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018200.hg.1 |
NM_206926.1 |
Refseq |