Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000525082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000525082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000525082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000525082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000525082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000526070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000529162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000531971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000531971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000531971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000531971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000533239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000533239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000533239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000533239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000534980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000617381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
ENST00000620157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_001257191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012495.hg.1 |
NM_004397.5 |
Refseq |