Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004914.hg.1 |
TC07000329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004914.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004914.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004914.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152687.hg.1 |
TC0700007610.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000254770.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000452107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000466041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000476479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
ENST00000486376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007610.hg.1 |
NM_018697.3 |
Refseq |