Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000215956.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000355257.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000355257.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000355257.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000355257.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000355257.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000355257.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000355257.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000401959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000402458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000402458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000402458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000463675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000463675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000463675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000463675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000469028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000469028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000469028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000469522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000469522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000469522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000488571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000488571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
ENST00000488571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_001003796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_001003796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_001003796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_001003796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_001003796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_001003796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_001003796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008837.hg.1 |
NM_005008.3 |
Refseq |