Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020441.hg.1 |
TC08001552.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020441.hg.1 |
ENST00000297350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020441.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178792.hg.1 |
TC0800011611.hg.1 |
Clariom D TC |