Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030565.hg.1 |
TC02001911.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030565.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030565.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193432.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193432.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193432.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193432.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193432.hg.1 |
TC0200012809.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000358912.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000466177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000468869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
ENST00000494203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012809.hg.1 |
NM_020651.3 |
Refseq |