Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022787.hg.1 |
TC06001876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022787.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022787.hg.1 |
ENST00000393019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022787.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022787.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022786.hg.1 |
TC06001876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022786.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022786.hg.1 |
ENST00000393019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022786.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022786.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022788.hg.1 |
TC06001876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022788.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022788.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022788.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012299.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184803.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184803.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184803.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184803.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184803.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184803.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184803.hg.1 |
TC0600012299.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600184805.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184805.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184805.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184805.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184805.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184805.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184805.hg.1 |
TC0600012299.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600184807.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184807.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184807.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184807.hg.1 |
ENST00000355773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184807.hg.1 |
ENST00000481996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184807.hg.1 |
NM_012434.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184807.hg.1 |
TC0600012299.hg.1 |
Clariom D TC |