Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134374.1 | NP_001127846.1 | 3 | 994 | 57 | |
NM_198572.2 | NP_940974.2 | 3 | 994 | 57 | ||
XM_011517021.1 | XP_011515323.1 | 3 | 994 | 57 | ||
XM_011517022.2 | XP_011515324.1 | 3 | 1280 | 57 | ||
XM_011517023.2 | XP_011515325.1 | 3 | 2298 | 57 | ||
XM_011517024.2 | XP_011515326.1 | 4 | 1519 | 57 | ||
XM_011517025.2 | XP_011515327.1 | 3 | 1954 | 57 | ||
XM_017013399.1 | XP_016868888.1 | 4 | 813 | 57 | ||
GenBank mRNA | AB092352.1 | - | 3 | 982 | 57 | |
AK301710.1 | - | 3 | 982 | 57 | ||
BC050390.1 | - | 3 | 984 | 5742710 | 57 | |
BC053547.1 | - | 3 | 969 | 5743874 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |