Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015044.4 | NP_055859.1 | 13 - 14 | 1372 | 93 | |
XM_017023074.1 | XP_016878563.1 | 14 - 15 | 5061 | 93 | ||
XM_017023075.1 | XP_016878564.1 | 13 - 14 | 1375 | 93 | ||
GenBank mRNA | AB029003.1 | - | 10 - 11 | 1002 | 93 | |
AF165531.1 | - | 13 - 14 | 1310 | 93 | ||
AF190863.1 | - | 13 - 14 | 1316 | 93 | ||
AF233522.1 | - | 13 - 14 | 1315 | 93 | ||
AF323754.1 | - | 14 - 15 | 1452 | 93 | ||
AK024619.1 | - | 13 - 14 | 1371 | 93 | ||
BC000284.1 | - | 13 - 14 | 1354 | 3356490 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |