Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001406.hg.1 |
TC02000132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001406.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001406.hg.1 |
ENST00000404924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001406.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001406.hg.1 |
ENST00000535786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001406.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001404.hg.1 |
TC02000132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001404.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001404.hg.1 |
ENST00000404924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001404.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001404.hg.1 |
ENST00000535786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001404.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006951.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000479859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147292.hg.1 |
TC0200006951.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000309033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000404924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000446425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
ENST00000535786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
NM_181713.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147296.hg.1 |
TC0200006951.hg.1 |
Clariom D TC |