Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024518.2 | NP_078794.1 | 2 - 3 | 432 | 88 | |
GenBank mRNA | AB052908.1 | - | 2 - 3 | 351 | 88 | |
AF304379.1 | - | 2 - 3 | 351 | 88 | ||
AF346596.1 | - | 2 - 3 | 327 | 88 | ||
AK297815.1 | - | 2 - 3 | 259 | 88 | ||
AK302076.1 | - | 2 - 3 | 406 | 88 | ||
AK315275.1 | - | 4 - 5 | 672 | 88 | ||
AY027538.1 | - | 2 - 3 | 321 | 88 | ||
BC098127.1 | - | 2 - 3 | 351 | 40003400 | 88 | |
CA488356.1 | - | 1 - 2 | 85 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |