Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012983.hg.1 |
ENST00000227667.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012983.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012983.hg.1 |
TC11001042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012982.hg.1 |
ENST00000227667.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012982.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012982.hg.1 |
TC11001042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012984.hg.1 |
ENST00000227667.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012984.hg.1 |
ENST00000375345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012984.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012984.hg.1 |
TC11001042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012986.hg.1 |
ENST00000227667.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012986.hg.1 |
ENST00000375345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012986.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012986.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012986.hg.1 |
TC11001042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012987.hg.1 |
ENST00000227667.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012987.hg.1 |
ENST00000375345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012987.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012987.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012987.hg.1 |
TC11001042.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167843.hg.1 |
TC1100009159.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000227667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000375345.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000433777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000470144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
ENST00000630701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
NM_000040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167842.hg.1 |
TC1100009159.hg.1 |
Clariom D TC |