Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016872.hg.1 |
TC0X001244.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016872.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016872.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171021.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171021.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171021.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171021.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171021.hg.1 |
TC0X00010431.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
ENST00000243298.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010431.hg.1 |
NM_016370.3 |
Refseq |