Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014047.hg.1 |
ENST00000252622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014047.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014047.hg.1 |
TC19001028.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000591745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
ENST00000591745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000591745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
ENST00000591745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009227.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
NM_016199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
TC1900009227.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000252622.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000585395.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000585409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000587502.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000589532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
ENST00000591515.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173593.hg.1 |
NM_016199.2 |
Refseq |