Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009182.hg.1 |
ENST00000351396.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009182.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009182.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009182.hg.1 |
TC10000772.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008769.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160021.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160021.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160021.hg.1 |
ENST00000369839.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160021.hg.1 |
NM_006951.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160021.hg.1 |
TC1000008769.hg.1 |
Clariom D TC |