Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006957.hg.1 |
TC03000327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006957.hg.1 |
ENST00000322241.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006957.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006956.hg.1 |
TC03000327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006956.hg.1 |
ENST00000322241.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006956.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153515.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153515.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153515.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153515.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153515.hg.1 |
TC0300007455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300153516.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153516.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153516.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153516.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153516.hg.1 |
TC0300007455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007455.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153517.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153517.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153517.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153517.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153517.hg.1 |
TC0300007455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300153518.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153518.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153518.hg.1 |
ENST00000322241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153518.hg.1 |
NM_080865.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153518.hg.1 |
TC0300007455.hg.1 |
Clariom D TC |