Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000588453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000588453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000590962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000590962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
ENST00000590962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160064.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160064.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160064.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160064.hg.1 |
TC1900008015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900160064.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160064.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160064.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000588453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000588453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000590962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000590962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
ENST00000590962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008015.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
NR_073032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
TC1900008015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000338502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000586301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000586958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000587013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
ENST00000587947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
NM_001042522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160065.hg.1 |
NR_073032.1 |
Refseq |