Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014990.hg.1 |
TC06001256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014990.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014990.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014990.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014991.hg.1 |
TC06001256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014991.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014991.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014991.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173136.hg.1 |
TC0600010757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173137.hg.1 |
TC0600010757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
ENST00000379660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_001142541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NM_015948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109913.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010757.hg.1 |
NR_109914.1 |
Refseq |