Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004615.3 | NP_004606.2 | 7 - 8 | 829 | 140 | |
GenBank mRNA | AK293178.1 | - | 8 - 9 | 814 | 140 | |
AK293534.1 | - | 9 - 10 | 983 | 140 | ||
AK293552.1 | - | 8 - 9 | 894 | 140 | ||
BC018036.1 | - | 7 - 8 | 788 | 4801388 | 140 | |
D10653.1 | - | 7 - 8 | 769 | 140 | ||
D29808.1 | - | 7 - 8 | 755 | 140 | ||
L10373.1 | - | 7 - 8 | 813 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |