Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006626.5 | NP_006617.1 | 1 - 2 | 70 | 72 | |
GenBank mRNA | AK292401.1 | - | 1 - 2 | 55 | 72 | |
BC037282.2 | - | 1 - 2 | 72 | 5266759 | 72 | |
BU600946.1 | - | 1 - 2 | 73 | 72 | ||
BX641237.1 | - | 1 - 2 | 71 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |