Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009605.hg.1 |
ENST00000262715.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009605.hg.1 |
ENST00000359243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009605.hg.1 |
ENST00000545983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009605.hg.1 |
ENST00000556935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009605.hg.1 |
NM_007110.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009605.hg.1 |
TC14000884.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009606.hg.1 |
ENST00000262715.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009606.hg.1 |
ENST00000359243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009606.hg.1 |
ENST00000556935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009606.hg.1 |
NM_007110.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009606.hg.1 |
TC14000884.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
ENST00000262715.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
ENST00000555008.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
ENST00000556935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
NM_007110.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
ENST00000262715.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
ENST00000555008.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
ENST00000556935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
NM_007110.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161669.hg.1 |
TC1400008588.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000262715.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000555008.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000555727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000556935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000557314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
NM_007110.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000262715.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000555008.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000555727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000556935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
ENST00000557314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
NM_007110.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161668.hg.1 |
TC1400008588.hg.1 |
Clariom D TC |