Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017022140.1 | XP_016877629.1 | 17 - 18 | 3904 | 60 | |
GenBank mRNA | AL833554.1 | - | 20 - 21 | 3932 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |