Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173483.3 | NP_775754.2 | 10 - 11 | 1303 | 80 | |
XM_011527692.2 | XP_011525994.1 | 11 - 12 | 1545 | 80 | ||
XM_011527693.2 | XP_011525995.1 | 10 - 11 | 1315 | 80 | ||
GenBank mRNA | AK096820.1 | - | 10 - 11 | 1303 | 80 | |
BC069351.1 | - | 9 - 10 | 1240 | 7262372 | 80 | |
BC093894.1 | - | 9 - 10 | 1177 | 7939739 | 80 | |
BC093896.1 | - | 9 - 10 | 1177 | 7939741 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection P450 Homo sapiens Probe spans exons |