Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007595.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007595.hg.1 |
ENST00000535781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007595.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007595.hg.1 |
TC20000531.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000156788.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156788.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000156788.hg.1 |
TC2000008104.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000156788.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156788.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
ENST00000266079.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008104.hg.1 |
NM_012469.3 |
Refseq |